Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncs1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328512.1 | NP_001315441.1 | 8 | 1840 | 147 | |
GenBank mRNA | BC055168.1 | - | 9 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |