Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhoub | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017784.3 | NP_001017784.2 | 3 | 2320 | 153 | |
GenBank mRNA | BC092896.1 | - | 4 | 1347 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |