Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gsta.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213394.1 | NP_998559.1 | 6 | 842 | 100 | |
XM_017358729.1 | XP_017214218.1 | 6 | 1035 | 100 | ||
GenBank mRNA | BC060914.1 | - | 6 | 842 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |