Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trim63a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002133.1 | NP_001002133.1 | 1 | 1449 | 91 | |
GenBank mRNA | BC071428.1 | - | 1 | 1449 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |