Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh1gc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012656.3 | NP_001012674.3 | 1 | 1454 | 66 | |
GenBank mRNA | AB160961.1 | - | 1 | 1430 | 66 | |
AB160962.1 | - | 1 | 1430 | 66 | ||
AB160963.1 | - | 1 | 1430 | 66 | ||
AB160964.1 | - | 1 | 1430 | 66 | ||
AY576955.1 | - | 1 | 1454 | 66 | ||
BC162469.1 | - | 1 | 1453 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |