Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pskh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020710.1 | NP_001018546.1 | 3 | 3147 | 102 | |
GenBank mRNA | BC095860.1 | - | 3 | 3147 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |