Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh1g26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024124.2 | NP_001019295.2 | 1 | 1153 | 99 | |
GenBank mRNA | AY576953.1 | - | 1 | 1153 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |