Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh2g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024176.2 | NP_001019347.1 | 1 | 942 | 84 | |
GenBank mRNA | AY583051.1 | - | 1 | 944 | 84 |
Gene Symbol: LOC103909937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328700.1 | NP_001315629.1 | 1 | 938 | 84 | |
GenBank mRNA | BC162556.1 | - | 1 | 938 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |