Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcdh2g7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024180.1 | NP_001019351.1 | 1 | 453 | 88 | |
GenBank mRNA | AY583056.1 | - | 1 | 453 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |