Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sox19a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130908.2 | NP_570983.2 | 2 | 1874 | 143 | |
GenBank mRNA | AB242331.1 | - | 2 | 1846 | 143 | |
BC078221.1 | - | 2 | 1815 | 143 | ||
EH609174.1 | - | 1 | 309 | 143 | ||
X79821.1 | - | 1746 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |