Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxg1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212914.1 | NP_998079.2 | 1 | 1950 | 109 | |
GenBank mRNA | BC067725.1 | - | 1 | 1950 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |