Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002575.1 | NP_001002575.1 | 6 - 7 | 558 | 97 | |
GenBank mRNA | BC076218.1 | - | 6 - 7 | 558 | 97 | |
BC092873.1 | - | 5 - 6 | 709 | 97 | ||
BC152265.1 | - | 6 - 7 | 577 | 97 | ||
BC153441.1 | - | 6 - 7 | 573 | 97 | ||
BC164974.1 | - | 5 - 6 | 490 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |