Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pde6d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002708.1 | NP_001002708.1 | 4 - 5 | 562 | 101 | |
GenBank mRNA | BC076096.1 | - | 4 - 5 | 562 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |