Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: impa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002745.1 | NP_001002745.1 | 8 - 9 | 969 | 124 | |
XM_005163396.4 | XP_005163453.1 | 8 - 9 | 986 | 124 | ||
XM_021479900.1 | XP_021335575.1 | 8 - 9 | 982 | 124 | ||
GenBank mRNA | BC076438.1 | - | 8 - 9 | 969 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |