Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpy19l1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002868.1 | NP_001002868.1 | 19 - 20 | 1917 | 86 | |
GenBank mRNA | AY648728.1 | - | 19 - 20 | 1917 | 86 | |
BC163516.1 | - | 19 - 20 | 1890 | 86 | ||
BC163534.1 | - | 19 - 20 | 1890 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |