Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh3bgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003524.2 | NP_001003524.1 | 10 - 11 | 680 | 71 | |
NM_001362477.1 | NP_001349406.1 | 9 - 10 | 656 | 71 | ||
NM_001362478.1 | NP_001349407.1 | 8 - 9 | 629 | 71 | ||
XM_005169682.4 | XP_005169739.1 | 11 - 12 | 849 | 71 | ||
XM_005169686.4 | XP_005169743.1 | 10 - 11 | 734 | 71 | ||
XM_005169688.4 | XP_005169745.1 | 9 - 10 | 587 | 71 | ||
XM_009291451.3 | XP_009289726.1 | 10 - 11 | 825 | 71 | ||
XM_021481385.1 | XP_021337060.1 | 8 - 9 | 562 | 71 | ||
GenBank mRNA | BC077137.1 | - | 10 - 11 | 653 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |