Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: suz12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003529.2 | NP_001003529.1 | 10 - 11 | 1328 | 126 | |
GenBank mRNA | BC078293.1 | - | 10 - 11 | 1331 | 126 | |
BC152625.1 | - | 10 - 11 | 1217 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |