Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: suz12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003529.2 | NP_001003529.1 | 11 - 12 | 1413 | 132 | |
GenBank mRNA | BC078293.1 | - | 11 - 12 | 1416 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |