Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003539.1 | NP_001003539.1 | 2 - 3 | 379 | 86 | |
GenBank mRNA | BC078187.1 | - | 2 - 3 | 379 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |