Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glrbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003587.1 | NP_001003587.1 | 3 - 4 | 523 | 115 | |
XM_021481003.1 | XP_021336678.1 | 3 - 4 | 536 | 115 | ||
GenBank mRNA | AB195560.1 | - | 2 - 3 | 223 | 115 | |
BC078269.1 | - | 3 - 4 | 523 | 115 | ||
KM655725.1 | - | 2 - 3 | 223 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |