Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ufc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003650.1 | NP_001003650.1 | 3 - 4 | 472 | 81 | |
GenBank mRNA | BC077079.1 | - | 3 - 4 | 472 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |