Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: thoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003758.1 | NP_001003758.1 | 2 - 3 | 349 | 71 | |
GenBank mRNA | BC079495.1 | - | 2 - 3 | 349 | 71 | |
BC152622.1 | - | 2 - 3 | 343 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |