Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: serpinf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004539.1 | NP_001004539.1 | 6 - 7 | 795 | 88 | |
GenBank mRNA | BC081582.1 | - | 6 - 7 | 795 | 88 | |
BC155160.1 | - | 6 - 7 | 801 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |