Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arrdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004561.1 | NP_001004561.1 | 1 - 2 | 203 | 147 | |
XM_005171783.4 | XP_005171840.1 | 1 - 2 | 188 | 147 | ||
GenBank mRNA | BC081631.1 | - | 1 - 2 | 157 | 147 | |
BC096962.1 | - | 1 - 2 | 182 | 147 | ||
BC152620.1 | - | 1 - 2 | 172 | 147 | ||
BC165815.1 | - | 1 - 2 | 151 | 147 | ||
CF416990.1 | - | 1 - 2 | 176 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |