Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: galca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005921.1 | NP_001005921.1 | 4 - 5 | 446 | 86 | |
GenBank mRNA | BC083517.1 | - | 4 - 5 | 446 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |