Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2k16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005963.2 | NP_001005963.2 | 6 - 7 | 1067 | 67 | |
GenBank mRNA | BC083304.1 | - | 6 - 7 | 1010 | 67 | |
BC165360.1 | - | 6 - 7 | 1032 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |