Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsd17b10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006098.1 | NP_001006098.1 | 4 - 5 | 554 | 70 | |
GenBank mRNA | BC083219.1 | - | 4 - 5 | 554 | 70 | |
BC165246.1 | - | 4 - 5 | 513 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |