Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acin1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005171275.1 | XP_005171332.1 | 12 - 13 | 4764 | 101 | |
XM_005171276.1 | XP_005171333.1 | 12 - 13 | 4761 | 101 | ||
GenBank mRNA | AI444258.1 | - | 2 - 3 | 239 | 101 | |
BC116536.1 | - | 6 - 7 | 637 | 101 | ||
CK238472.1 | - | 1 - 2 | 179 | 101 | ||
CT659964.2 | - | 5 - 6 | 637 | 101 | ||
EB785743.1 | - | 3 - 4 | 447 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |