Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpkow | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007207.1 | NP_001007208.1 | 8 - 9 | 1258 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |