Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpkow | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007207.1 | NP_001007208.1 | 9 - 10 | 1319 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |