Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npy7r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007218.1 | NP_001007219.1 | 1 | 651 | 69 | |
XM_021481005.1 | XP_021336680.1 | 2 | 1135 | 69 | ||
GenBank mRNA | BC163317.1 | - | 1 | 675 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |