Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oxct1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007291.1 | NP_001007292.1 | 10 - 11 | 1075 | 56 | |
GenBank mRNA | BC078408.2 | - | 10 - 11 | 1075 | 56 | |
BC163972.1 | - | 10 - 11 | 1102 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |