Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: neil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007335.1 | NP_001007336.1 | 5 - 6 | 736 | 88 | |
XM_005157318.3 | XP_005157375.1 | 5 - 6 | 795 | 88 | ||
GenBank mRNA | BC085550.1 | - | 5 - 6 | 736 | 88 | |
BC155157.1 | - | 5 - 6 | 716 | 88 | ||
BC164271.1 | - | 5 - 6 | 668 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |