Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftr72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007387.2 | NP_001007388.2 | 3 - 4 | 755 | 75 | |
GenBank mRNA | BC085564.1 | - | 3 - 4 | 755 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |