Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007412.1 | NP_001007413.1 | 3 - 4 | 504 | 96 | |
GenBank mRNA | BC085373.1 | - | 3 - 4 | 504 | 96 | |
BC165511.1 | - | 3 - 4 | 283 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |