Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007447.1 | NP_001007448.1 | 4 - 5 | 613 | 104 | |
GenBank mRNA | BC085392.1 | - | 4 - 5 | 613 | 104 | |
BC164562.1 | - | 3 - 4 | 408 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |