Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007770.1 | NP_001007771.1 | 1 - 2 | 180 | 85 | |
GenBank mRNA | BC085569.1 | - | 1 - 2 | 180 | 85 | |
BC164956.1 | - | 1 - 2 | 201 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |