Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007770.1 | NP_001007771.1 | 3 - 4 | 346 | 122 | |
GenBank mRNA | BC085569.1 | - | 3 - 4 | 346 | 122 | |
BC164956.1 | - | 3 - 4 | 367 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |