Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008576.2 | NP_001008576.1 | 5 - 6 | 1128 | 88 | |
XM_017356481.2 | XP_017211970.1 | 5 - 6 | 980 | 88 | ||
XM_021476852.1 | XP_021332527.1 | 5 - 6 | 1175 | 88 | ||
XM_021476853.1 | XP_021332528.1 | 5 - 6 | 938 | 88 | ||
GenBank mRNA | BC086960.1 | - | 5 - 6 | 1128 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |