Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfap2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008576.2 | NP_001008576.1 | 7 | 3686 | 167 | |
GenBank mRNA | BC086960.1 | - | 7 | 3727 | 167 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |