Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008628.1 | NP_001008628.1 | 2 - 3 | 745 | 67 | |
GenBank mRNA | BC086729.1 | - | 2 - 3 | 745 | 67 | |
BC165767.1 | - | 2 - 3 | 667 | 67 | ||
BC169189.1 | - | 728 | 67 | |||
FJ032033.1 | - | 2 - 3 | 633 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |