Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012374.1 | NP_001012374.1 | 3 - 4 | 1167 | 76 | |
GenBank mRNA | AJ879586.1 | - | 3 - 4 | 1167 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Danio rerio Probe spans exons |