Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itga6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013448.1 | NP_001013466.1 | 2 - 3 | 355 | 75 | |
XM_009291986.3 | XP_009290261.1 | 2 - 3 | 389 | 75 | ||
GenBank mRNA | BC090458.1 | - | 2 - 3 | 355 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |