Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mybpc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013511.2 | NP_001013529.2 | 21 - 22 | 2274 | 109 | |
XM_017353828.2 | XP_017209317.1 | 20 - 21 | 2339 | 109 | ||
XM_021470159.1 | XP_021325834.1 | 21 - 22 | 2308 | 109 | ||
XM_021470160.1 | XP_021325835.1 | 20 - 21 | 2336 | 109 | ||
XM_021470161.1 | XP_021325836.1 | 20 - 21 | 2286 | 109 | ||
XM_021470162.1 | XP_021325837.1 | 20 - 21 | 2223 | 109 | ||
GenBank mRNA | BC091662.1 | - | 2268 | 109 | ||
EE714901.1 | - | 5 - 6 | 632 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |