Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013573.1 | NP_001013591.1 | 3 - 4 | 405 | 112 | |
XM_021481013.1 | XP_021336688.1 | 3 - 4 | 432 | 112 | ||
XM_021481015.1 | XP_021336690.1 | 3 - 4 | 432 | 112 | ||
XM_021481016.1 | XP_021336691.1 | 3 - 4 | 454 | 112 | ||
GenBank mRNA | BC090463.1 | - | 3 - 4 | 405 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |