Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trim2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014371.1 | NP_001014393.1 | 4 - 5 | 737 | 68 | |
XM_005170889.4 | XP_005170946.1 | 4 - 5 | 816 | 68 | ||
GenBank mRNA | BC091789.1 | - | 4 - 5 | 737 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |