Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: actr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017542.2 | NP_001017542.1 | 1 - 2 | 124 | 62 | |
XM_005169510.4 | XP_005169567.1 | 1 - 2 | 149 | 62 | ||
XM_021480582.1 | XP_021336257.1 | 1 - 2 | 43 | 62 | ||
GenBank mRNA | BC092187.1 | - | 1 - 2 | 128 | 62 | |
BC153558.1 | - | 1 - 2 | 48 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |