Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alkbh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017609.1 | NP_001017609.1 | 3 - 4 | 688 | 116 | |
GenBank mRNA | BC093288.1 | - | 3 - 4 | 688 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |