Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhbdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017614.1 | NP_001017614.1 | 5 - 6 | 815 | 69 | |
XM_021481406.1 | XP_021337081.1 | 5 - 6 | 860 | 69 | ||
GenBank mRNA | BC092835.1 | - | 5 - 6 | 815 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |