Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: praf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017698.1 | NP_001017698.1 | 1 - 2 | 182 | 65 | |
GenBank mRNA | BC093309.1 | - | 1 - 2 | 182 | 65 | |
BC164952.1 | - | 1 - 2 | 209 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |